Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Jars, with screw cap

35 ml, 38 mm, 48 mm, White
  SEMA3715
 :   SEMADENI
Jars, with screw cap
  SEMA3715
 :  SEMADENI
Jars, with screw cap
Jars, with screw cap

35 ml, 38 mm, 48 mm, White

 
46,10 €
   
1 * 100 ST
 

PP, white.

  • Good chemical resistance
  • Good temperature resistance

Ideal for collecting, storing and transporting samples.

 
  • Ext.Ø:
    38 mm
  • Capacity:
    35 ml
  • Colour:
    White
  • Height:
    48 mm
  • Seq:
    03