Boot covers, Secutex® per, PP/PE
Elasticated, White, U*
Catalog # 113-0779
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Jump to:
Product Details & Documents
These boot covers provide protection against fine dusts and splashes of aggressive liquid chemicals. Partial resistance to splashing.
- Smooth, lint-free and abrasion-resistant material (PP spunbond with PE laminate)
- Permeable to air and water vapour
- Impervious to particles, with partial resistance to splashing and also anti-static
- Choice of boot covers either elasticated or with ties
Ideal for use in the remediation of contaminated sites, building reconstruction or in asbestos abatement as well as in spray work in agriculture or in industry and car paint shops.
*One size fits all.
Certifications: Cat. III, type 5 + 6
Specifications
- Shoe size:U*
- Colour:White
- Description:Elasticated
- Cat. No.:113-0779
- Garment application:Chemical protection|Laboratory|Industrial