Safety spectacles, XC™
IR, anti-scratch, Blue, IR Shade 5 PC
Catalog # DALL1011020
Supplier: HONEYWELL SAFETY
Jump to:
Product Details & Documents
The large curved lens of these safety glasses offers a total wrap-around coverage and provides exceptional peripheral vision along with complete protection.
- Adjustable arm length and lens angle
- Pads for excellent non slip function
- Extra flexible frame; for men and women
- Multi-materials provide extra comfort; soft brow, nose bridge and temples
EN 166, EN 170
: EN 166, EN 170
Specifications
- Type:IR, anti-scratch
- Colour:Blue
- Seq:15
- Lenses:IR Shade 5 PC
Accessories
Product Information
- Catalog #AvailabilityPrice
- Adapter corrective lenses for safety spectacles, XC™Adapter corrective lenses for safety spectacles, XC™111-3720Log in for availability1 * 1 ST - 50,70 €