Order Entry
Germany
Orders LinkContactUsLinkComponent
Storage Bins

Storage Bins

Blue, 345×213×200 mm, PP
Catalog # SEMA2797
Supplier:   SEMADENI
Storage Bins
Catalog # SEMA2797
Supplier:  SEMADENI
Storage Bins
Storage Bins

Blue, 345×213×200 mm, PP

Price
25,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Made of PP.

  • Secure stacking
  • Made of high quality material and are dimensionally stable
  • Supporting edge and smooth inner surfaces for easy cleaning

Storage bins with large labelling field for content description.

Specifications
  • Colour:
    Blue
  • Material:
    PP
  • Seq:
    04
  • W×D×H:
    345×213×200 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.