Order Entry
Germany
Orders LinkContactUsLinkComponent
Cyclopentane, Supelco®

Cyclopentane, Supelco®

Glass bottle, 5 mL
Catalog # 29680-5ML
Flammable
Supplier:   Merck
Cyclopentane, Supelco®
Catalog # 29680-5ML
Supplier:  Merck
CAS Number:  287-92-3
Cyclopentane, Supelco®
Cyclopentane, Supelco®

Glass bottle, 5 mL

CAS Number:  287-92-3
Price
31,10 €
Log in for availability
1 * 5 mL
Product Details & Documents
Synonyms:  Pentamethylene
  • CAS Number:287-92-3
  • Formula:C₅H₁₀
  • MDL Number:MFCD00001356
  • Molecular Weight:70.13 g/mol
  • Merck Index:13,02769
  • Boiling Pt:49 °C (1013 hPa)
  • Melting Pt:–93.3 °C
  • Density:0.746 g/cm³ (20 °C)
  • Flash Pt:–41 °C
  • Storage Temperature:Ambient
  • UN:1146
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:206-016-6
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.