Order Entry
Germany
Orders LinkContactUsLinkComponent
D-(+)-Galactose, Millipore®

D-(+)-Galactose, Millipore®

Glass bottle for solids, 500 g
Catalog # SIAL48260-500G-F
Supplier:   Merck
D-(+)-Galactose, Millipore®
Catalog # SIAL48260-500G-F
Supplier:  Merck
CAS Number:  59-23-4
D-(+)-Galactose, Millipore®
D-(+)-Galactose, Millipore®

Glass bottle for solids, 500 g

CAS Number:  59-23-4
Price
367,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04356
  • Boiling Pt:527 °C (1013 hPa)
  • Melting Pt:163…169 °C
  • Density:1.5 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:200-416-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications