Order Entry
Germany
Orders LinkContactUsLinkComponent
Methylcyclohexane, Supelco®

Methylcyclohexane, Supelco®

Glass bottle, 1 mL
Catalog # 66294-1ML
Flammable
Health
Irritant
Environment
Supplier:   Merck
Methylcyclohexane, Supelco®
Catalog # 66294-1ML
Supplier:  Merck
CAS Number:  108-87-2
Methylcyclohexane, Supelco®
Methylcyclohexane, Supelco®

Glass bottle, 1 mL

CAS Number:  108-87-2
Price
78,10 €
Log in for availability
1 * 1 mL
Product Details & Documents
  • CAS Number:108-87-2
  • Formula:C₆H₁₁CH₃
  • MDL Number:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:13,06074
  • Boiling Pt:101 °C (1013 hPa)
  • Melting Pt:–126 °C
  • Density:0.7693 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • UN:2296
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-624-3

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL

Specifications