Order Entry
Germany
Orders LinkContactUsLinkComponent
D-(+)-Mannose, Millipore®

D-(+)-Mannose, Millipore®

Glass bottle for solids, 100 g
Artikelnummer SIAL63580-100G
Hersteller:   Merck
D-(+)-Mannose, Millipore®
Artikelnummer SIAL63580-100G
Hersteller:  Merck
CAS-Nummer:  3458-28-4
D-(+)-Mannose, Millipore®

Glass bottle for solids, 100 g

CAS-Nummer:  3458-28-4
Preis
212,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,05772
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:133 °C
  • Density:1.539 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:222-392-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.