Order Entry
Germany
Orders LinkContactUsLinkComponent
Diethyl ether, Supelco®

Diethyl ether, Supelco®

Glass bottle, 10 mL
Catalog # 91238-10ML-F
Flammable
Irritant
Supplier:   Merck
Diethyl ether, Supelco®
Catalog # 91238-10ML-F
Supplier:  Merck
CAS Number:  60-29-7
Diethyl ether, Supelco®
Diethyl ether, Supelco®

Glass bottle, 10 mL

CAS Number:  60-29-7
Price
351,00 €
Log in for availability
1 * 10 mL
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • UN:1155
  • ADR:3,I
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.