Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), 1
Catalog # UPTIUP86510C
Supplier: Uptima
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # UPTIUP86510C
Supplier: Uptima
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1
- Description:TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)



