Streptavidin
Glass bottle for solids, 10 mg
Catalog # UPTIUP51558E
Supplier: Uptima
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Cat. No.:UPTIUP51558E
- Protein/peptide name:Streptavidin
- Storage conditions:Freezer
- MDL No.:MFCD00082035
- CAS No.:9013-20-1
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



