Tween® 20 (Polysorbate)
Glass bottle, 10 mL
Catalog # UPTIUP158741
Supplier: Uptima
Jump to:
Product Details & Documents
Synonyms: Polyoxyethylene sorbitan monolaurate
- CAS Number:9005-64-5
- Formula:C₅₈H₁₁₄O₂₆
- MDL Number:MFCD00165986
- Molecular Weight:1227.53 g/mol
- Density:1.1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:500-018-3
Specifications
Specifications
- Pack type:Glass bottle
- Pk:10 mL



