D-(+)-Biotin
Glass bottle for solids, 200 mg
Catalog # UPTIUP10685D
Supplier: Uptima
Jump to:
Product Details & Documents
Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Number:58-85-5
- Formula:C₁₀H₁₆N₂O₃S
- MDL Number:MFCD00005541
- Molecular Weight:244.31 g/mol
- Merck Index:13,01231
- Melting Pt:232…233 °C
- Density:1.41 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:200-399-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:200 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



