Brij® 35
1
Catalog # UPTIUP091870
Supplier: Uptima
Jump to:
Product Details & Documents
Synonyms: Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
- CAS Number:9002-92-0
- Formula:C₅₈H₁₁₈O₂₄
- MDL Number:MFCD00080891
- Molecular Weight:1199.56 g/mol
- Storage Temperature:Ambient
- EINECS:500-002-6
Specifications
Specifications
- Pk:1



