Order Entry
Germany
Orders LinkContactUsLinkComponent
Saccharometers | Avantor

Saccharometers

60 - 80, 1,0, 200 mm
Catalog # AMARH846468
Supplier:   Amarell
Saccharometers
Catalog # AMARH846468
Supplier:  Amarell
Saccharometers
Saccharometers

60 - 80, 1,0, 200 mm

Price
33,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Saccharometers with reference temperature of 20 °C.

Specifications
  • Length:
    200 mm
  • Seq:
    06
  • Division:
    1,0
  • Range:
    60 - 80
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.