Orders LinkContactUsLinkComponent
Safety spectacles, A800
Safety spectacles, A800
Anti-scratch, Grey, Clear PC
Catalog # 111-0166
Supplier:   HONEYWELL SAFETY
Safety spectacles, A800
Catalog # 111-0166
Supplier:  HONEYWELL SAFETY
Safety spectacles, A800
Safety spectacles, A800
Anti-scratch, Grey, Clear PC
Price
6,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Single lens safety glasses for general protection against impact.
  • Temples with soft and padded inserts with extra flex built in
  • Slim profile for smaller faces
  • Sporty styling provides a fashionable fit
Suitable for applications in utilities, manufacturing, biotech, laboratories, cleanrooms, ship building and automotive manufacturing.
 : EN 166.
Specifications
  • Type:
    Anti-scratch
  • Colour:
    Grey
  • Seq:
    01
  • Lenses:
    Clear PC