Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Capillary columns, VF-17ms

Capillary, VF-17ms, 0,15 µm, 0,15 mm, 20 m
  VARICP5884
 :   VARIAN
Capillary columns, VF-17ms
  VARICP5884
 :  VARIAN
Capillary columns, VF-17ms
Capillary columns, VF-17ms

Capillary, VF-17ms, 0,15 µm, 0,15 mm, 20 m

 
756,00 €
   
1 * 1 ST
 

VF-17ms is a 50% phenyl, 50% dimethylpolysiloxane, medium polarity, low bleed column for increased sensitivity, accuracy and instrument uptime.

  • Deactivation technology improves data quality
  • Ideal EPA confirmation column for ultimate confidence
  • Bonded and cross-linked to allow solvent rinsing, reducing replacement costs
  • Improves column stability
 
  • Column Format:
    Capillary
  • Film thickness:
    0,15 µm
  • Ø int.:
    0,15 mm
  • Length:
    20 m
  • Seq:
    09
  • Phase:
    VF-17ms