Operating supplies
Pump tubing, SIPS
Catalog # VARI9910075900
Supplier: VARIAN
Jump to:
Product Details & Documents
The operating supplies set includes two 500 ml constant pressure bottle, a diluent bottle (1 L), a three-way SIPS tee piece assembly for SIPS-20, six SIPS pump tubings, a SIPS kit and ten pack SIPS pump bands.
Specifications
- Model:SIPS
- Description:Pump tubing
- Seq:02