Decontamination Solution, DNA/RNA-ExitusPlus™
DNA/RNA-ExitusPlus™, refill pack, 1 L
Supplier: PanReac AppliChem
Decontamination Solution, DNA/RNA-ExitusPlus™
Catalog # A7089.1000RF
Supplier: PanReac AppliChem
CAS Number:
Jump to:
Product Details & Documents
Solution for the removal of DNA and RNA contamination from surfaces. Suitable for cleaning PCR work stations and equipment, electrophoresis equipment, as well as pipettes and reaction tubes, for example.
- All components are readily bio-degradable and not harmful or toxic for humans
- Doesn't contain aggressive mineral acids or alkaline substances
- No organic solvents or volatile components
- Delivered as spray bottles or refill bottles (RF)
Documents
Certificates
Search by Lot NumberApplichem Lifescience Effective DNA and RNA Decontamination in One DNA RNA-ExitusPlusTM Applichem Lifescience ExitusPlus™ – Effective Solution – Effective Decontamination Applichem Lifescience Products for the Investigation of COVID-19 Applichem Lifescience Products Supporting your Work Against COVID-19
Product Literature
Nucleic Acid Decontamination with DNA-ExitusPlus™ Technology Nucleic acid decontamination with DNA-ExitusPlus
Nucleic Acid Decontamination with DNA RNA-ExitusPlusTM
Additional Product Details
TDS DNA and RNA ExitusPlus™
Datasheet
Specifications
Specifications
- Pk:1 L
- Description:DNA/RNA-ExitusPlus™, refill pack
- Cat. No.:A7089.1000RF
- Storage temperature:Ambient
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



