Orders LinkContactUsLinkComponent
Screw caps, PP
Screw caps, PP
with Butyl/PTFE seal, 38 mm, Black
Catalog # 215-2377
Supplier:   GRAVIS
Screw caps, PP
Catalog # 215-2377
Supplier:  GRAVIS
Screw caps, PP
Screw caps, PP
with Butyl/PTFE seal, 38 mm, Black
Price
276,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Specifications
  • Thread:
    38 mm
  • Colour:
    Black
  • Description:
    with Butyl/PTFE seal