2-(2-Bromophenyl)ethanol 99%
Glass bottle for solids, 5 g
- :1074-16-4
- :C₈H₉BrO
- :MFCD00093566
- :201.06 g/mol
- :97 °C (.7 mmHg)
- :>110 °C
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5755 to 1.5775 (20°C, 589 nm)
- GC:≥98.5 %
- Pack type:Glass bottle for solids
- Pk:5 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



