2-(2-Bromophenyl)ethanol 99%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
2-(2-Bromophenyl)ethanol 99%
Catalog # ACRO350550010
Supplier: Thermo Fisher Scientific
CAS Number: 1074-16-4
Jump to:
Product Details & Documents
- CAS Number:1074-16-4
- Formula:C₈H₉BrO
- MDL Number:MFCD00093566
- Molecular Weight:201.06 g/mol
- Boiling Pt:97 °C (.7 mmHg)
- Flash Pt:>110 °C
Documents
ACRO35055
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5755 to 1.5775 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



