Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Water for analysis

Plastic bottle, 2,5 L
Catalog # ACRO389400025
   Safer Human and Environmental Health
Water for analysis
Catalog # ACRO389400025
   Safer Human and Environmental Health
CAS Number:  7732-18-5
Water for analysis
Water for analysis

Plastic bottle, 2,5 L

CAS Number:  7732-18-5
Price
44,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (760 mmHg)
  • Melting Pt:0 °C
  • Density:1 g/cm³
  • Storage Temperature:Ambient
  • EINECS:231-791-2

 

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Non-volatile matter:
    ≤0.0005 %
  • Residue after ign. (sulfated):
    ≤0.0002 %
  • pH:
    5 to 8
  • Absorbance:
    ≤0.01 (water, 1 cm, 254 nm)
  • Chloride (Cl):
    ≤0.1 ppm
  • Nitrate (NO3):
    ≤0.2 ppm
  • Phosphate (PO4):
    ≤0.05 ppm
  • Sulfate (SO4):
    ≤1 ppm
  • Silicate (as SiO2):
    ≤0.01 ppm
  • Aluminium (Al):
    ≤0.02 ppm
  • Ammonium (NH4):
    ≤0.01 ppm
  • Antimony (Sb):
    ≤0.01 ppm
  • Barium (Ba):
    ≤0.01 ppm
  • Bismuth (Bi):
    ≤0.01 ppm
  • Boron (B):
    ≤0.01 ppm
  • Cadmium (Cd):
    ≤0.01 ppm
  • Calcium (Ca):
    ≤0.1 ppm
  • Chromium (Cr):
    ≤0.01 ppm
  • Cobalt (Co):
    ≤0.01 ppm
  • Copper (Cu):
    ≤0.01 ppm
  • Gold (Au):
    ≤0.01 ppm
  • Iron (Fe):
    ≤0.01 ppm
  • Lithium (Li):
    ≤0.02 ppm
  • Magnesium (Mg):
    ≤0.05 ppm
  • Manganese (Mn):
    ≤0.01 ppm
  • Mercury (Hg):
    ≤0.05 ppm
  • Molybdenum (Mo):
    ≤0.01 ppm
  • Nickel (Ni):
    ≤0.01 ppm
  • Potassium (K):
    ≤0.05 ppm
  • Selenium (Se):
    ≤0.01 ppm
  • Silver (Ag):
    ≤0.01 ppm
  • Sodium (Na):
    ≤0.1 ppm
  • Strontium (Sr):
    ≤0.05 ppm
  • Tin (Sn):
    ≤0.01 ppm
  • Titanium (Ti):
    ≤0.01 ppm
  • Vanadium (V):
    ≤0.01 ppm
  • Zinc (Zn):
    ≤0.05 ppm
  • Zirconium (Zr):
    ≤0.01 ppm
  • Heavy metals (as Pb):
    ≤0.01 ppm
  • KMnO4-reducing substance:
    Passes test
  • Conductivity:
    ≤6 µS/cm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Cat. No.:
    ACRO389400025
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications