Orders LinkContactUsLinkComponent
DNAOUT™ DNA Removal Solution
DNA OUT™, refill, 1 L
Catalog # 786-75
Supplier:   G-Biosciences
DNAOUT™ DNA Removal Solution
Catalog # 786-75
Supplier:  G-Biosciences
CAS Number:  
DNAOUT™ DNA Removal Solution
DNAOUT™ DNA Removal Solution
DNA OUT™, refill, 1 L
CAS Number:  
Price
113,00 €
Log in for availability
1 * 1 L
Product Details & Documents
DNAOUT™ is a DNA removal solution for the rapid decontamination of work areas from contaminating DNA. Simply spray the work area and wait 10-15 seconds and then rinse with Molecular Grade Water.
  • Ready-to-use with convenient spray bottle and refill bottles available
  • Remove contaminating DNA prior to sensitive work, including PCR
  • Removes amplicon, plasmid and genomic DNA
  • Fast acting (10-15 seconds)
  • Ideal for cleaning centrifuge tubes, reaction tubes, PCR machine surface, pipettes, lab benches, lab equipment and more
Specifications
Specifications
  • Pk:
    1 L
  • Description:
    DNA OUT™, refill
Specifications