Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium azide solution 1%

Sodium azide solution 1%

Glass bottle, 100 mL
Catalog # 786-299
Supplier:   G-Biosciences
Sodium azide solution 1%
Catalog # 786-299
Supplier:  G-Biosciences
CAS Number:  26628-22-8
Sodium azide solution 1%

Glass bottle, 100 mL

CAS Number:  26628-22-8
Price
36,70 €
Log in for availability
1 * 100 mL
Product Details & Documents

G-Biocide-III is sodium azide based preservative. Azide anions bind to trivalent iron in porphyrin complexes and inhibit the activity of catalase, peroxidases and cytochrome oxidases. It acts as bacteriostatic by inhibiting cytochrome oxidase.

  • CAS number: 26628-22-8 Molecular formula: NaN3 Molecular weight: 65.0099 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N Recommended concentration for use: 0.02 to 0.05%

A 1% aqueous solution of sodium azide [154 mm] for use as a preservative for laboratory reagents. The use of the 1% solution is convenient and minimizes exposure and contamination risks normally encountered while handling sodium azide powder. Used as preservative to preserve wide range of research reagents such as buffers, blocking solutions for ELISA, chromatography resins and Immunoglobulins storage solutions.

Synonyms:  Sodium triazide
  • CAS Number:26628-22-8
  • Formula:NaN₃
  • MDL Number:MFCD00003536
  • Molecular Weight:65.01 g/mol

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.