Order Entry
Germany
Orders LinkContactUsLinkComponent
Dry buffer packs, JAW™

Dry buffer packs, JAW™

Tris buffered saline packs, JAW™, 1X, 1 l/pack, 25mM Tris, 140mM sodium chloride and 3mM potassium chloride, 7,25 - 7,55, 20
Catalog # 786-288
Supplier:   G-Biosciences
Dry buffer packs, JAW™
Catalog # 786-288
Supplier:  G-Biosciences
CAS Number:  
Dry buffer packs, JAW™

Tris buffered saline packs, JAW™, 1X, 1 l/pack, 25mM Tris, 140mM sodium chloride and 3mM potassium chloride, 7,25 - 7,55, 20

CAS Number:  
Price
130,00 €
Log in for availability
1 * 20 ST
Product Details & Documents

Premixed buffer powders that simply require adding water to generate a ready-to-use buffer.

Documents

Specifications

Specifications

  • Pk:
    20
  • Description:
    Tris buffered saline packs, JAW™, 1X, 1 l/pack
  • pH value:
    7,25 - 7,55
  • Contents:
    25mM Tris, 140mM sodium chloride and 3mM potassium chloride

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.