Orders LinkContactUsLinkComponent
Gel loading dyes, Glow™
Gel loading dyes, Glow™
Glow™ dye 6X, CyanoBlue™, 1,5 mL
Catalog # 786-104
Supplier:   G-Biosciences
Gel loading dyes, Glow™
Catalog # 786-104
Supplier:  G-Biosciences
CAS Number:  
Gel loading dyes, Glow™
Gel loading dyes, Glow™
Glow™ dye 6X, CyanoBlue™, 1,5 mL
CAS Number:  
Price
58,40 €
Log in for availability
1 * 1,5 mL
Product Details & Documents
These premixed and ready-to-use gel dyes are designed to minimise the use of ethidium bromide. Glow dyes provide intense DNA/RNA bands with little background or band distortion on any type of agarose or acrylamide gel.
  • Dyes for DNA are Ficoll®-based
  • Glow loading dyes already contain ethidium bromide
  • Both RNA loading dyes are formamide-based
  • Glow RNA for quick screenings of RNA preps
Specifically formulated for nucleic acid electrophoresis.
Specifications
Specifications
  • Pk:
    1,5 mL
  • Description:
    Glow™ dye 6X, CyanoBlue™