J.T.Baker® DILUT-IT™, Buffer Solutions, Concentrate
DILUT-IT™ concentrate, 6,00, Citrate, 1
Catalog # 4854.
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Buffer concentrate (citrate/sodium hydroxide), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 6.00 ± 0.02 (20°C) Titrisol® 1 * 1 Ampoul Catalog #1.09886.0001
Product 1.09886.0001 is similar to 4854. but manufacturer and specifications may vary.
Buffer solution pH 6 AVS TITRINORM Plastic bottle 1L 1 * 1 L Catalog #32036.294
Product 32036.294 is similar to 4854. but manufacturer and specifications may vary.
Product Details & Documents
Volumetric concentrate, for dilution to 500 ml.
- Traceable to NIST
Documents
Specifications
Specifications
- pH buffer type:Citrate
- Pk:1
- Description:DILUT-IT™ concentrate
- pH value:6,00



