Order Entry
Germany
Orders LinkContactUsLinkComponent
J.T.Baker® DILUT-IT™, Buffer Solutions, Concentrate

J.T.Baker® DILUT-IT™, Buffer Solutions, Concentrate

DILUT-IT™ concentrate, 2,00, ±0,02, Citrate, 20 °C, 1
Catalog # 4851.
Supplier:   Avantor
J.T.Baker® DILUT-IT™, Buffer Solutions, Concentrate
Catalog # 4851.
Supplier:  Avantor
CAS Number:  
J.T.Baker® DILUT-IT™, Buffer Solutions, Concentrate

DILUT-IT™ concentrate, 2,00, ±0,02, Citrate, 20 °C, 1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Volumetric concentrate, for dilution to 500 ml.

  • Traceable to NIST

Documents

Specifications

Specifications

  • pH buffer type:
    Citrate
  • Pk:
    1
  • Reference temperature:
    20 °C
  • Description:
    DILUT-IT™ concentrate
  • pH accuracy:
    ±0,02
  • pH value:
    2,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.