Order Entry
Germany
Orders LinkContactUsLinkComponent
Bottle Carriers

Bottle Carriers

Bottle carriers, Red, Up to six bottles, 95 mm
Catalog # KART395
Supplier:   KARTELL
Bottle Carriers
Catalog # KART395
Supplier:  KARTELL
Bottle Carriers
Bottle Carriers

Bottle carriers, Red, Up to six bottles, 95 mm

Price
131,00 €
Log in for availability
1 * 5 ST
Product Details & Documents

PP, coloured.

  • Ergonomic design of the handle
  • Can be easily stacked
  • Very good chemical and temperature resistance

Designed to carry 6 and 4 bottles, with maximum diameter up to 90 mm and 110 mm respectively.

Specifications
  • For:
    Up to six bottles
  • Colour:
    Red
  • Description:
    Bottle carriers
  • Container max. Ø:
    95 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.