Orders LinkContactUsLinkComponent
Adapters, reduction, Preciso
Adapters, reduction, Preciso
Reduction, 45/40 NS, 34/35
Catalog # WITG0.500.020
Supplier:   witeg Labortechnik
Adapters, reduction, Preciso
Catalog # WITG0.500.020
Supplier:  witeg Labortechnik
Adapters, reduction, Preciso
Adapters, reduction, Preciso
Reduction, 45/40 NS, 34/35
Price
49,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
Certifications: According to DIN 12257.
Specifications
  • Cone size:
    45/40 NS
  • Socket:
    34/35
  • Type:
    Reduction
  • Seq:
    20