Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Tissue processing/embedding cassettes, Swingsette®, M515

Purple
  720-1628
Tissue processing/embedding cassettes, Swingsette®, M515
  720-1628
Tissue processing/embedding cassettes, Swingsette®, M515
Tissue processing/embedding cassettes, Swingsette®, M515

Purple

 
275,00 €
   
1 * 1.500 ST
 

Acetal polymer.

  • Integral, hinged lid, with large tab for easy opening
  • Disposable plastic cassettes highly resistant to histological solvents
  • Flow-through slots maximise fluid exchange
  • Front face inclined 45° to enable use with leading automatic labelling systems.

 : Each case contains three boxes of 500 cassettes.

 
  • Colour:
    Purple