Orders LinkContactUsLinkComponent
Abbe refractometers, NAR
Abbe refractometers, NAR
NAR-1T (liquids), 0 − 95 °Bx, 1,30 − 1,70 nD
Catalog # ATAG1211
Supplier:   Atago
Abbe refractometers, NAR
Catalog # ATAG1211
Supplier:  Atago
Abbe refractometers, NAR
Abbe refractometers, NAR
NAR-1T (liquids), 0 − 95 °Bx, 1,30 − 1,70 nD
Price
3 320,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The NAR Abbe refractometers can meet many of the requirements for routine measurements.
Single wavelengths and standard temperatures can be measured at extended ranges, as well as elevated temperatures, measurements of solid samples (films, plastic sheets).
Specifications
  • Model:
    NAR-1T (liquids)
  • Range:
    0 − 95 °Bx
  • Refractive index:
    1,30 − 1,70 nD
  • Temperature measurement:
    5...50 °C
  • Cat. No.:
    ATAG1211
  • Light source:
    LED
  • Temperature sensor:
    ±0,2 °C
  • Temperature sensor accuracy:
    ±0,2 °C
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Secondary prism for NAR-1T•LO
    Secondary prism for NAR-1T•LO
    ATAGRE-1111
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Thermo display unit (DTM-1) for NAR-1T•LO, NAR-2T•LO and NAR-2T•HI
    Thermo display unit (DTM-1) for NAR-1T•LO, NAR-2T•LO and NAR-2T•HI
    ATAGRE-1165
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Lamp cable for NAR-1T•LO
    Lamp cable for NAR-1T•LO
    ATAGRE-1175
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Secondary prism for NAR-3T
    Secondary prism for NAR-3T
    ATAGRE-1204
    Log in for availability
    1 * 1 ST