Order Entry
Germany
Orders LinkContactUsLinkComponent
ICP-MS interference check mixes

ICP-MS interference check mixes

6020 Interference check solution A. For use with ICP-MS systems. Contains: 20000 ppm Cl, 3000 ppm Ca, 2500 ppm Fe and Na, 2000 ppm C, 1000 ppm Al, Mg, P, K, S and 20 ppm Mo and Ti. 100 ml, 5% HNO₃, 100 mL
Catalog # HEWL5188-6526
Supplier:   Agilent
ICP-MS interference check mixes
Catalog # HEWL5188-6526
Supplier:  Agilent
CAS Number:  
ICP-MS interference check mixes
ICP-MS interference check mixes

6020 Interference check solution A. For use with ICP-MS systems. Contains: 20000 ppm Cl, 3000 ppm Ca, 2500 ppm Fe and Na, 2000 ppm C, 1000 ppm Al, Mg, P, K, S and 20 ppm Mo and Ti. 100 ml, 5% HNO₃, 100 mL

CAS Number:  
Price
817,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

It offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for atomic spectroscopy.

  • It is works in AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations

Standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Description:
    6020 Interference check solution A. For use with ICP-MS systems. Contains: 20000 ppm Cl, 3000 ppm Ca, 2500 ppm Fe and Na, 2000 ppm C, 1000 ppm Al, Mg, P, K, S and 20 ppm Mo and Ti. 100 ml

Specifications