Order Entry
Germany
Orders LinkContactUsLinkComponent
Corning® Matrigel® Basement Membrane Matrix, Corning | Avantor

Corning® Matrigel® Basement Membrane Matrix, Corning

Matrigel® Matrix, growth factor reduced, high concentration, 10 mL
Catalog # 734-1441
   Low Manufacturing Impact
Supplier:   Corning
Corning® Matrigel® Basement Membrane Matrix, Corning
Catalog # 734-1441
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
Corning® Matrigel® Basement Membrane Matrix, Corning
Corning® Matrigel® Basement Membrane Matrix, Corning

Matrigel® Matrix, growth factor reduced, high concentration, 10 mL

Wechseln zu:
CAS Number:  
Price
811,15 €
Anmelden für die Verfügbarkeit
1 * 10 mL
Product Details & Documents

Growth Factor Reduced (GFR) formulation is well suited for applications that require a more highly defined basement membrane preparation.

  • Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
  • Extensive PCR testing is performed to screen for a number of pathogens
  • Tested and found negative for bacteria, fungi, and mycoplasma
  • Protein concentrations determined by Lowry method
  • Endotoxin units measured by Limulus Amoebocyte Lysate assay
  • Matrigel Matrix gel stability tested for a period of 14 days at 37 °C
  • Biological activity determined for each lot using a neurite outgrowth assay.

Corning Matrigel matrix is a solubilised basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Application
• Cell Growth and Differentiation
• Metabolism/Toxicology Studies
• Invasion Assays
In Vitro and In Vivo Angiogenesis Assays
In Vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice

Preparation and Storage
Store at –20 °C. Avoid multiple freeze-thaws.

Note: For research use only. Not intended for diagnostic or therapeutic procedures.

When warmed, the solution polymerizes to produce a biologically active matrix material resembling the mammalian cellular basement membrane. It is ideal for use with in vitro cell cultures of a broad range of cell types, including both primary and stem cell applications.

Spezifikationen:

Spezifikationen:

  • Pk:
    10 mL
  • Description:
    Matrigel® Matrix, growth factor reduced, high concentration
  • Cat. No.:
    734-1441
  • Environmental certification:
    ISO 14001 - Bedford
  • Environmentally Preferable:
    Low Manufacturing Impact

Spezifikationen:

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.