DIAMANTPASTE 1µM
Catalog # PLANG3401
Supplier: PLANO W. PLANNET
Frequently Bought Together
FAM Benzine DIN 51635, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Petroleum spirit, 100…120 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
GLYCEROL PURISS., MEETS ANALYTICAL SPECI 1 * 1 L
Supplier: Merck
Tetrahydrofuran, anhydrous 99.9% stabilised, extra pure
Supplier: Thermo Fisher Scientific
n-Pentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Methylbutane for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol, extra pure
Supplier: Bernd Kraft
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Videojet Technologies, OPLOSMIDDEL 750 M
Supplier: MARKET SOURCE PART PROCESS
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Methyl ethyl ketone ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals