Stands for MSC-Advantage
Fixed height floor stand, 750 mm working height, for 1,2 m cabinet
Catalog # 135-6832
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Specifications
- Description:Fixed height floor stand, 750 mm working height, for 1,2 m cabinet
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



