Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Decontamination Solution, DNA/RNA-ExitusPlus™

DNA/RNA-ExitusPlus™, refill pack, 2,5 L
Catalog # A7089.2500RF
Supplier:   PanReac AppliChem
Decontamination Solution, DNA/RNA-ExitusPlus™
Catalog # A7089.2500RF
Supplier:  PanReac AppliChem
CAS Number:  
Decontamination Solution, DNA/RNA-ExitusPlus™
Decontamination Solution, DNA/RNA-ExitusPlus™

DNA/RNA-ExitusPlus™, refill pack, 2,5 L

CAS Number:  
Price
280,16 €
Log in for availability
1 * 2,5 L
Product Details & Documents

Solution for the removal of DNA and RNA contamination from surfaces. Suitable for cleaning PCR work stations and equipment, electrophoresis equipment, as well as pipettes and reaction tubes, for example.

  • All components are readily bio-degradable and not harmful or toxic for humans
  • Doesn't contain aggressive mineral acids or alkaline substances
  • No organic solvents or volatile components
  • Delivered as spray bottles or refill bottles (RF)
Specifications

Specifications

  • Pk:
    2,5 L
  • Description:
    DNA/RNA-ExitusPlus™, refill pack
  • Cat. No.:
    A7089.2500RF
  • Storage temperature:
    Ambient

Specifications