Safety Containers
with handle, 7 l, 178×240 mm
Catalog # DELT240007
Supplier: DELTALAB
Jump to:
Product Details & Documents
Safety containers made of either PP or PE.
- Suitable for solid waste type II and III, enable incineration
- Container body consists hazard warning label
It consists two lids, an entire opening, taking advantage of the total diameter of the mouth of the container, allows to throw away bigger volume waste. A partial opening, which may be closed temporarily or definitively. It embodies two devices in order to easy the extraction and disconnection of dental and analytical needles.
Certifications: In accordance with European standards ONU ADR, ISO 6706, BS 5404.
Specifications
- Capacity:7 l
- Description:with handle
- Seq:04
- Ø ext.×H:178×240 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



