Tripod stands
120×250 mm
Catalog # 451-0160
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Tripod stands made of stainless steel or zinc plated steel.
Please note: Zinc plating of tripod stands serves as temporary corrosion protection. May peel off after direct contact with open burner flame.
Specifications
- Int.Ø×H:120×250 mm
- Seq:03