Order Entry
Germany
Orders LinkContactUsLinkComponent
VWR®, Cover Slips, Rectangular

VWR®, Cover Slips, Rectangular

24×60 mm, #1
Catalog # 631-1575
Supplier:   VWR Collection
VWR®, Cover Slips, Rectangular
Catalog # 631-1575
Supplier:  VWR Collection
VWR®, Cover Slips, Rectangular
VWR®, Cover Slips, Rectangular

24×60 mm, #1

Price
88,90 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

These cover slips are specially designed for diagnostic applications.

  • Made of pure white Hydrolytic Class 1 glass

Thickness: 0.13 - 0.16 mm

Thickness: 0,13 - 0,16 mm

 : In dispenser boxes with separate lid.

Specifications
  • No.:
    #1
  • L×W:
    24×60 mm
  • Seq:
    10