Order Entry
Germany
Orders LinkContactUsLinkComponent
Distillation apparatus, micro

Distillation apparatus, micro

Receiver for short path distillation, with 4 test tubes 3 ml:0,5 ml
Catalog # WITG2.996.005
Supplier:   witeg Labortechnik
Distillation apparatus, micro
Catalog # WITG2.996.005
Supplier:  witeg Labortechnik
Distillation apparatus, micro
Distillation apparatus, micro

Receiver for short path distillation, with 4 test tubes 3 ml:0,5 ml

Price
82,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Short path distilling apparatus, micro, complete.

Specifications
  • Description:
    Receiver for short path distillation, with 4 test tubes 3 ml:0,5 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.