Orders LinkContactUsLinkComponent
Distillation apparatus, micro
Distillation apparatus, micro
Distillation apparatus, short path, micro
Catalog # WITG2.996.000
Supplier:   witeg Labortechnik
Distillation apparatus, micro
Catalog # WITG2.996.000
Supplier:  witeg Labortechnik
Distillation apparatus, micro
Distillation apparatus, micro
Distillation apparatus, short path, micro
Price
286,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Short path distilling apparatus, micro, complete.
Specifications
  • Description:
    Distillation apparatus, short path, micro