Order Entry
Germany
Orders LinkContactUsLinkComponent
 

VWR® Storage Vials

8,00 ml, 16,6×61 mm, Clear, No
Catalog # 548-0821
Supplier:   VWR Collection
VWR® Storage Vials
Catalog # 548-0821
Supplier:  VWR Collection
VWR® Storage Vials
VWR® Storage Vials

8,00 ml, 16,6×61 mm, Clear, No

Price
443,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Screw neck vials in clear or amber glass, hydrolytic class 1, for sample storage. A broad range of closed top screw caps are available with different types of septa.

Specifications
  • Cap included:
    No
  • Ø×H:
    16,6×61 mm
  • Capacity:
    8,00 ml
  • Colour:
    Clear
  • Seq:
    02