Order Entry
Germany
Orders LinkContactUsLinkComponent
Process pH combination electrodes

Process pH combination electrodes

pH combination electrode, with dome membrane, H 8381 HD, 2 - 13, 0...+100 °C, Glass, Referid®, KPG annular gap, 12×320 mm, Screw plug head
Catalog # SCOI285111023
Supplier:   SI Analytics
Process pH combination electrodes
Catalog # SCOI285111023
Supplier:  SI Analytics
Process pH combination electrodes
Process pH combination electrodes

pH combination electrode, with dome membrane, H 8381 HD, 2 - 13, 0...+100 °C, Glass, Referid®, KPG annular gap, 12×320 mm, Screw plug head

Price
408,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Connection:
    Screw plug head
  • Ø×L:
    12×320 mm
  • Type:
    H 8381 HD
  • Description:
    pH combination electrode, with dome membrane
  • Material:
    Glass
  • Seq:
    26
  • Diaphr.:
    KPG annular gap
  • Electrolyte:
    Referid®
  • Temp.:
    0...+100 °C
  • pH:
    2 - 13
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.