Order Entry
Germany
Orders LinkContactUsLinkComponent
Jerricans, polyethlene

Jerricans, polyethlene

10 l, 298×198×264 mm, Black
Catalog # DUEH5.09.010
Supplier:   DUPERTHAL
Jerricans, polyethlene
Catalog # DUEH5.09.010
Supplier:  DUPERTHAL
Jerricans, polyethlene
Jerricans, polyethlene

10 l, 298×198×264 mm, Black

Price
30,05 €
Log in for availability
1 * 1 ST
Product Details & Documents

PE, electrostatically conductive.

  • Good chemical resistance
  • Good temperature resistance
  • Recyclable
Specifications
  • Capacity:
    10 l
  • Colour:
    Black
  • W×D×H:
    298×198×264 mm
Accessories

Product Information

  • Catalog #
    For
    Ø
    Availability
    Price
  • Safety funnel
    Safety funnel
    DUEH5.08.001
    Flammable liquids
    120 mm
    Log in for availability
    1 * 1 ST - 185,07 €

Product Information

  • Catalog #
    Availability
    Price
  • Special pliers
    Special pliers
    DUEH5.09.003
    Log in for availability
    1 * 1 ST - 8,05 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.