Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Single Hexagonal Rotor (For 90 ea. 1.5 ml microcentrifuge tubes) 1 * 1 items
Catalog # LBCP7816400
Supplier:
LABCONCO
Expand image
Single Hexagonal Rotor (For 90 ea. 1.5 ml microcentrifuge tubes) 1 * 1 items
Catalog # LBCP7816400
Supplier:
LABCONCO