Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Extension rods, with M10 thread

12 mm, 500 mm, M10
Catalog # USBE2161
Supplier:   USBECK Laborgeräte
Extension rods, with M10 thread
Catalog # USBE2161
Extension rods, with M10 thread
Extension rods, with M10 thread

12 mm, 500 mm, M10

Price
21,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Extension rods made from stainless steel.

  • One internal and one external thread M 10
  • Ø 12 mm
  • Size of jaw 10 mm
Specifications
  • Ø:
    12 mm
  • Thread:
    M10
  • Length:
    500 mm