Order Entry
Germany
Orders LinkContactUsLinkComponent
Extension rods, with M10 thread

Extension rods, with M10 thread

12 mm, 250 mm, M10
Catalog # USBE2159
Supplier:   USBECK Laborgeräte
Extension rods, with M10 thread
Catalog # USBE2159
Extension rods, with M10 thread
Extension rods, with M10 thread

12 mm, 250 mm, M10

Price
17,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Extension rods made from stainless steel.

  • One internal and one external thread M 10
  • Ø 12 mm
  • Size of jaw 10 mm
Specifications
  • Ø:
    12 mm
  • Thread:
    M10
  • Length:
    250 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.