Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Pharmaceutical Refrigerator MED-340, 330 l, +2 to +20 C 1 * 1 items
Catalog # KIRSMED340
Supplier:
KIRSCH PHILIPP
Expand image
Pharmaceutical Refrigerator MED-340, 330 l, +2 to +20 C 1 * 1 items
Catalog # KIRSMED340
Supplier:
KIRSCH PHILIPP